Anti-Clostridium botulinum A Toxoid Mouse Monoclonal Antibody [clone: 9L109]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC5851-40HEA 4410 DKK
USBIC5851-40H
Anti-Clostridium botulinum A Toxoid Mouse Monoclonal Antibody [clone: 9L109]
Antistoffer
Type: Primary
Antigen: Clostridium botulinum A Toxoid
Clonality: Monoclonal
Clone: 9L109
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR