Anti-Clostridium botulinum A Toxoid Mouse Monoclonal Antibody [clone: 7C219]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC5851-40BEA 3290 DKK
USBIC5851-40B
Anti-Clostridium botulinum A Toxoid Mouse Monoclonal Antibody [clone: 7C219]
Antistoffer
Recommended dilution: Immunofluorescence: 1:50

Type: Primary
Antigen: Clostridium botulinum A Toxoid
Clonality: Monoclonal
Clone: 7C219
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR