Anti-CLOCK Sheep Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC5843-16EA 4780 DKK
USBIC5843-16
Anti-CLOCK Sheep Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 0.5 ?g/ml

Type: Primary
Antigen: CLOCK (clock circadian regulator)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR