Anti-CLOCK Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC5843-01-100EA 4650 DKK
USBIC5843-01-100
Anti-CLOCK Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: CLOCK (clock circadian regulator)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Drosophila
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR