Anti-CLIC5 Mouse Monoclonal Antibody [clone: 1E6]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC5840-46AEA 4790 DKK
USBIC5840-46A
Anti-CLIC5 Mouse Monoclonal Antibody [clone: 1E6]
Antistoffer
Recommended dilution: Immunochemistry: 10 ?g/ml; Western Blot: 1:500-1:1000

Type: Primary
Antigen: CLIC5 (chloride intracellular channel 5)
Clonality: Monoclonal
Clone: 1000000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR