Anti-CLIC4 Goat Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC5840-45EEA 4730 DKK
USBIC5840-45E
Anti-CLIC4 Goat Polyclonal Antibody
Antistoffer
Recommended dilution: Immunofluorescence: 5 ?g/ml; ELISA: 1:64000; Western Blot: 0.1-0. 3?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: CLIC4 (chloride intracellular channel 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Goat
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR