Anti-CLIC4 Goat Polyclonal Antibody
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBIC5840-45BEA 4120 DKK
USBIC5840-45B
Anti-CLIC4 Goat Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:64000
Type: Primary
Antigen: CLIC4 (chloride intracellular channel 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestil nu