Anti-CLEC5A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 11C8]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC5839-75EA 4500 DKK
USBIC5839-75
Anti-CLEC5A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 11C8]
Antistoffer
Do not freeze!

Type: Primary
Antigen: CLEC5A (C-type lectin domain family 5, member A)
Clonality: Monoclonal
Clone: 11C8
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR