Anti-CLEC16A Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC5839-74BEA 3700 DKK
USBIC5839-74B
Anti-CLEC16A Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: CLEC16A (C-type lectin domain family 16, member A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR