Anti-CLEC14A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 12K67]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC5839-73TEA 5030 DKK
USBIC5839-73T
Anti-CLEC14A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 12K67]
Antistoffer
Do not freeze!

Type: Primary
Antigen: CLEC14A (C-type lectin domain family 14, member A)
Clonality: Monoclonal
Clone: 12K67
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR