Anti-p90 RSK1 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI168415EA 6290 DKK
USBI168415
Anti-p90 RSK1 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:2000-1:5000; Western Blot: 1:200-1:1000

Type: Primary
Antigen: p90 RSK1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR