Anti-FCGR3A Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 13H55]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI137641EA 4190 DKK
USBI137641
Anti-FCGR3A Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 13H55]
Antistoffer
Do not freeze!

Type: Primary
Antigen: FCGR3A (Fc fragment of IgG, low affinity IIIa, receptor (CD16a))
Clonality: monoclonal
Clone: 13H55
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR