Anti-SP3 Mouse Monoclonal Antibody [clone: 3H7]
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBI133736EA 3600 DKK
USBI133736
Anti-SP3 Mouse Monoclonal Antibody [clone: 3H7]
Antistoffer
Recommended dilution: Immunofluorescence: 10 ?g/ml
Type: Primary
Antigen: SP3 (Sp3 transcription factor)
Clonality: monoclonal
Clone: 3H7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human