Anti-SGCA Rabbit Polyclonal Antibody (Biotin)
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBI041647-BIOTINEA 6360 DKK
USBI041647-BIOTIN
Anti-SGCA Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: SGCA (sarcoglycan, alpha (50kDa dystrophin-associated glycoprotein))
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human