Anti-MTRF1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Leverandør: US Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBI038684-APEA 6350 DKK
USBI038684-AP
Anti-MTRF1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: MTRF1 (mitochondrial translational release factor 1)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human