Anti-MTL5 Rabbit Polyclonal Antibody
Leverandør: US Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBI038661EA 4380 DKK
USBI038661
Anti-MTL5 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: MTL5 (metallothionein-like 5, testis-specific (tesmin))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human