Anti-MTHFD2L Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Leverandør: US Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBI038655-FITCEA 6350 DKK
USBI038655-FITC
Anti-MTHFD2L Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: MTHFD2L (methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2-like)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human