Anti-MT-CO3 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Leverandør: US Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBI038647-APCEA 6350 DKK
USBI038647-APC
Anti-MT-CO3 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antistoffer
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: MT-CO3 (mitochondrially encoded cytochrome c oxidase III)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human