Anti-MST1R Rabbit Polyclonal Antibody
Leverandør: US Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBI038642EA 4380 DKK
USBI038642
Anti-MST1R Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 1:10-50
Type: Primary
Antigen: MST1R (macrophage stimulating 1 receptor (c-met-related tyrosine kinase))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse