Anti-MS4A14 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverandør: US Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI038619-HRPEA 6360 DKK
USBI038619-HRP
Anti-MS4A14 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: MS4A14 (membrane-spanning 4-domains, subfamily A, member 14)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR