Anti-MRPS24 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Leverandør: US Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI038610-APEA 6360 DKK
USBI038610-AP
Anti-MRPS24 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50
Do not freeze!

Type: Primary
Antigen: MRPS24 (mitochondrial ribosomal protein S24)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR