Anti-MRPL46 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Leverandør: US Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBI038599-PEEA 6360 DKK
USBI038599-PE
Anti-MRPL46 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antistoffer
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: MRPL46 (mitochondrial ribosomal protein L46)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human