Anti-MRPL28 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Leverandør: US Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI038593-APCEA 6420 DKK
USBI038593-APC
Anti-MRPL28 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antistoffer
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: MRPL28 (mitochondrial ribosomal protein L28)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR