Anti-MPHOSPH6 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Leverandør: US Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBI038558-HRPEA 6350 DKK
USBI038558-HRP
Anti-MPHOSPH6 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: MPHOSPH6 (M-phase phosphoprotein 6)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human