Anti-MOAP1 Rabbit Polyclonal Antibody (Biotin)

Leverandør: US Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI038523-BIOTINEA 6350 DKK
USBI038523-BIOTIN
Anti-MOAP1 Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: MOAP1 (modulator of apoptosis 1)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR