Anti-MMS19 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Leverandør: US Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI038516-APEA 6350 DKK
USBI038516-AP
Anti-MMS19 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: MMS19 (MMS19 nucleotide excision repair homolog (S. cerevisiae))
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR