Anti-MLL2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Leverandør: US Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBI038475-FITCEA 6420 DKK
USBI038475-FITC
Anti-MLL2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
Recommended dilution: FLISA: 1:1000
Do not freeze!
Type: Primary
Antigen: MLL2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human