Anti-MLKL Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Leverandør: US Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBI038472-APCEA 6500 DKK
USBI038472-APC
Anti-MLKL Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antistoffer
Recommended dilution: Western Blot: 1:100-1:500
Do not freeze!
Type: Primary
Antigen: MLKL (mixed lineage kinase domain-like)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse