Anti-MLH1 Rabbit Polyclonal Antibody (Biotin)

Leverandør: US Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI038468-BIOTINEA 6350 DKK
USBI038468-BIOTIN
Anti-MLH1 Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50

Type: Primary
Antigen: MLH1 (mutL homolog 1)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR