Anti-MKKS Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Leverandør: US Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI038450-APCEA 6350 DKK
USBI038450-APC
Anti-MKKS Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antistoffer
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: MKKS (McKusick-Kaufman syndrome)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR