Anti-MGLL Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverandør: US Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI038416-FITCEA 6350 DKK
USBI038416-FITC
Anti-MGLL Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: MGLL (monoglyceride lipase)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR