Anti-MFS10 Rabbit Polyclonal Antibody (Biotin)
Leverandør: US Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBI038401-BIOTINEA 6350 DKK
USBI038401-BIOTIN
Anti-MFS10 Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: MFS10
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human