Anti-GRM1 Rabbit Polyclonal Antibody

Leverandør: US Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI038373EA 4680 DKK
USBI038373
Anti-GRM1 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50

Type: Primary
Antigen: GRM1 (glutamate receptor, metabotropic 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR