Anti-MESP1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Leverandør: US Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI038364-PEEA 6350 DKK
USBI038364-PE
Anti-MESP1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antistoffer
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: MESP1 (mesoderm posterior 1 homolog (mouse))
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR