Anti-MESDC2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverandør: US Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI038363-HRPEA 6420 DKK
USBI038363-HRP
Anti-MESDC2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50; Immunofluorescence: 1:10-50

Type: Primary
Antigen: MESDC2 (mesoderm development candidate 2)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR