Anti-MEPE Rabbit Polyclonal Antibody (Biotin)
Leverandør: US Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBI038359-BIOTINEA 6350 DKK
USBI038359-BIOTIN
Anti-MEPE Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: MEPE (matrix extracellular phosphoglycoprotein)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human