Anti-MAP3K4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverandør: US Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI038350-HRPEA 6350 DKK
USBI038350-HRP
Anti-MAP3K4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50

Type: Primary
Antigen: MAP3K4 (mitogen-activated protein kinase kinase kinase 4)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR