Anti-VIM Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI031444EA 3230 DKK
USBI031444
Anti-VIM Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Immunofluorescence: 1:5000; Western Blot: 1:10000

Type: Primary
Antigen: VIM (vimentin)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human, Mammalian, Mouse, Pig, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR