Anti-UB Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI031437EA 3230 DKK
USBI031437
Anti-UB Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Immunofluorescence: 1:500-1:1000; Western Blot: 1:5000-1:10000

Type: Primary
Antigen: UB (ubiquitin)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR