Anti-PRPH Rabbit Polyclonal Antibody
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBI031421EA 3780 DKK
USBI031421
Anti-PRPH Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: PRPH (peripherin)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat