Anti-Neurofilament L, 68kD Mouse Monoclonal Antibody [clone: 13B841]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI031403EA 3230 DKK
USBI031403
Anti-Neurofilament L, 68kD Mouse Monoclonal Antibody [clone: 13B841]
Antistoffer
Recommended dilution: Immunofluorescence: 1:100-1:500; Western Blot: 1:5000-1:10000

Type: Primary
Antigen: Neurofilament L, 68kD
Clonality: Monoclonal
Clone: 13B841
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Avian, Bovine, Human, Mammalian, Mouse, Pig, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR