Anti-GAPDH Mouse Monoclonal Antibody [clone: 13B822]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI031365EA 3000 DKK
USBI031365
Anti-GAPDH Mouse Monoclonal Antibody [clone: 13B822]
Antistoffer
Recommended dilution: Immunofluorescence: 1:100; Western Blot: 1:1000

Type: Primary
Antigen: GAPDH (glyceraldehyde-3-phosphate dehydrogenase)
Clonality: Monoclonal
Clone: 13B822
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM Kappa
Reactivity: Bovine, Human, Mouse, Pig, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR