Anti-FBL Mouse Monoclonal Antibody [clone: 13B818]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI031359EA 3280 DKK
USBI031359
Anti-FBL Mouse Monoclonal Antibody [clone: 13B818]
Antistoffer
Recommended dilution: Immunofluorescence: 1:1000-1:5000

Type: Primary
Antigen: FBL (fibrillarin)
Clonality: Monoclonal
Clone: 13B818
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: C. elegans, Drosophila, Human, Rat, Yeast
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR