Anti-Cherry Mouse Monoclonal Antibody [clone: 13B813]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI031353EA 3230 DKK
USBI031353
Anti-Cherry Mouse Monoclonal Antibody [clone: 13B813]
Antistoffer
Recommended dilution: Immunofluorescence: 1:500; Western Blot: 1:2000

Type: Tag
Antigen: Cherry
Clonality: Monoclonal
Clone: 13B813
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR