Anti-ZNF622 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 208CT34.1.1]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI031343-PEEA 5310 DKK
USBI031343-PE
Anti-ZNF622 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 208CT34.1.1]
Antistoffer
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze! Light sensitive

Type: Primary
Antigen: ZNF622 (zinc finger protein 622)
Clonality: Monoclonal
Clone: 208CT34.1.1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR