Anti-YEATS4 Mouse Monoclonal Antibody [clone: 204CT4.7.2]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI031338EA 3940 DKK
USBI031338
Anti-YEATS4 Mouse Monoclonal Antibody [clone: 204CT4.7.2]
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-16000

Type: Primary
Antigen: YEATS4 (YEATS domain containing 4)
Clonality: Monoclonal
Clone: 204CT4.7.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR