Anti-UBTD1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 189CT14.5.4]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI031318-FITCEA 5310 DKK
USBI031318-FITC
Anti-UBTD1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 189CT14.5.4]
Antistoffer
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: UBTD1 (ubiquitin domain containing 1)
Clonality: Monoclonal
Clone: 189CT14.5.4
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR