Anti-TSC2 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 249CT19.1.3]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI031313-FITCEA 5310 DKK
USBI031313-FITC
Anti-TSC2 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 249CT19.1.3]
Antistoffer
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100

Type: Primary
Antigen: TSC2 (tuberous sclerosis 2)
Clonality: Monoclonal
Clone: 249CT19.1.3
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR