Anti-TLE1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 381CT18.5.1]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI031293-APCEA 5310 DKK
USBI031293-APC
Anti-TLE1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 381CT18.5.1]
Antistoffer
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-250

Type: Primary
Antigen: TLE1 (transducin-like enhancer of split 1 (E(sp1) homolog, Drosophila))
Clonality: Monoclonal
Clone: 381CT18.5.1
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR