Anti-THOC1 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 301CT40.5.1]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI031290-APEA 5310 DKK
USBI031290-AP
Anti-THOC1 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 301CT40.5.1]
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-250

Type: Primary
Antigen: THOC1 (THO complex 1)
Clonality: Monoclonal
Clone: 301CT40.5.1
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Mouse
Isotype: IgG3 Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR